Assay Details
| Gene Symbol: MIR99A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_035386.1 | 1 | 47 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC106508546 | LOC106508546 | Sense | Chr. 13 - 180777580 - 181371532 on NCBI Build Sscrofa11.1 | ||
| Downstream | microRNA let-7c | MIRLET7C |
MIRNLET7C, ssc-let-7c |
647.0 | Sense | Chr. 13 - 181303209 - 181303302 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR99A
Genomic Map