Assay Details
Gene Symbol: MIR193A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038540.1 | 1 | 40 | 57 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-365-2 | MIR365-2 | ssc-mir-365-2 | -12.2kb | Sense | Chr. 12 - 43324725 - 43324804 on NCBI Build Sscrofa11.1 |
Downstream | RAB11 family interacting protein 4 | RAB11FIP4 | 23.7kb | Sense | Chr. 12 - 43360758 - 43474235 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
