Assay Details
Gene Symbol: MIR7857 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128470.1 | 1 | 46 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger protein 184 | ZNF184 | -24.5kb | Anti-sense | Chr. 7 - 21413273 - 21435236 on NCBI Build Sscrofa11.1 | |
Downstream | histone H2B type 1-H-like | LOC110261671 | 122.3kb | Anti-sense | Chr. 7 - 21582043 - 21582522 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
