Assay Details
Gene Symbol: MIR9798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128480.1 | 1 | 45 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | R-spondin 4 | RSPO4 | -30.7kb | Anti-sense | Chr. 17 - 34268702 - 34310073 on NCBI Build Sscrofa11.1 | |
Downstream | angiopoietin 4 | ANGPT4 | 5.1kb | Anti-sense | Chr. 17 - 34345926 - 34394977 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
