Assay Details
Gene Symbol: MIR9801 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128483.1 | 1 | 39 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | olfactomedin 2 | OLFM2 | -34.3kb | Sense | Chr. 2 - 68734078 - 68807207 on NCBI Build Sscrofa11.1 | |
Downstream | retinol dehydrogenase 8 | RDH8 | 30.2kb | Anti-sense | Chr. 2 - 68871707 - 68878222 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
