Assay Details
Gene Symbol: MIR9807 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128492.1 | 1 | 37 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | CD83 molecule | CD83 | -373.2kb | Anti-sense | Chr. 7 - 10346122 - 10385968 on NCBI Build Sscrofa11.1 | |
Downstream | jumonji and AT-rich interaction domain containing 2 | JARID2 | 599.0kb | Anti-sense | Chr. 7 - 11358243 - 11601715 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
