Assay Details
Gene Symbol: MIR9838 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128525.1 | 1 | 34 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cyclin D1 binding protein 1 | CCNDBP1 | -35.8kb | Anti-sense | Chr. 1 - 128253111 - 128269534 on NCBI Build Sscrofa11.1 | |
Downstream | ubiquitin protein ligase E3 component n-recognin 1 | UBR1 | 13.9kb | Sense | Chr. 1 - 128319311 - 128474330 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
