Assay Details
Gene Symbol: MIR9843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128530.1 | 1 | 41 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | sphingomyelin synthase 2 | SGMS2 | Sense | Chr. 8 - 114052726 - 114145946 on NCBI Build Sscrofa11.1 | ||
Downstream | 3'-phosphoadenosine 5'-phosphosulfate synthase 1 | PAPSS1 | 141.3kb | Anti-sense | Chr. 8 - 114252059 - 114369309 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
