Assay Details
Gene Symbol: MIR9858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128554.1 | 1 | 34 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | carbonic anhydrase 2 | CA2 | -147.0 | Anti-sense | Chr. 4 - 51194686 - 51211436 on NCBI Build Sscrofa11.1 | |
Downstream | carbonic anhydrase 3 | CA3 | 15.8kb | Anti-sense | Chr. 4 - 51227485 - 51237861 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
