Assay Details
Gene Symbol: MIR296 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037200.1 | 1 | 38 | 48 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | aminopeptidase like 1 | NPEPL1 | -79.1kb | Anti-sense | Chr. 17 - 58874499 - 58891249 on NCBI Build Sscrofa11.1 | |
Downstream | GNAS complex locus | GNAS |
NESP55, PORGSA1, SCG6 |
15.2kb | Anti-sense | Chr. 17 - 58985580 - 59053022 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
