Assay Details
| Gene Symbol: MIR362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037201.1 | 1 | 35 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-500 | MIR500 |
MIR500A, ssc-mir-500 |
-393.0 | Sense | Chr. X - 43711254 - 43711333 on NCBI Build Sscrofa11.1 |
| Downstream | chloride voltage-gated channel 5 | CLCN5 | CLC-5 | 67.2kb | Sense | Chr. X - 43779016 - 43809472 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR362
Genomic Map