Assay Details
Gene Symbol: MIR205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038496.1 | 1 | 32 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | calcium/calmodulin dependent protein kinase IG | CAMK1G | -136.6kb | Sense | Chr. 9 - 133351523 - 133380454 on NCBI Build Sscrofa11.1 | |
Downstream | plexin A2 | PLXNA2 | 1.1mb | Anti-sense | Chr. 9 - 134584760 - 134800471 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
