Assay Details
| Gene Symbol: MIR27A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038498.1 | 1 | 42 | 60 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-23a | MIR23A | ssc-mir-23a | -108.0 | Sense | Chr. 2 - 65308117 - 65308186 on NCBI Build Sscrofa11.1 |
| Downstream | microRNA mir-24-1 | MIR24-1 |
MIR24, ssc-mir-24, ssc-mir-24-1 |
78.0 | Sense | Chr. 2 - 65308453 - 65308524 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR27A
Genomic Map