Assay Details
Gene Symbol: MIR129B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038601.1 | 1 | 34 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | leptin | LEP |
OB, OBS |
-33.1kb | Sense | Chr. 18 - 20106867 - 20124071 on NCBI Build Sscrofa11.1 |
Downstream | staphylococcal nuclease and tudor domain containing 1 | SND1 | 104.2kb | Sense | Chr. 18 - 20261473 - 20653621 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
