Assay Details
Gene Symbol: MIR187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038602.2 | 1 | 40 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | polypeptide N-acetylgalactosaminyltransferase 1 | GALNT1 | GalNAc-T1 | -181.0kb | Anti-sense | Chr. 6 - 119295896 - 119493901 on NCBI Build Sscrofa11.1 |
Downstream | chromosome 6 C18orf21 homolog | C6H18orf21 | 60.4kb | Anti-sense | Chr. 6 - 119735388 - 119745729 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
