Assay Details
Gene Symbol: MIR219A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038605.1 | 1 | 42 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | hydroxysteroid 17-beta dehydrogenase 8 | HSD17B8 |
FABGL, HKE6, KE6, Ke-6 |
-1.1kb | Sense | Chr. 7 - 25247577 - 25249786 on NCBI Build Sscrofa11.1 |
Downstream | ring finger protein 1 | RING1 | 586.0 | Sense | Chr. 7 - 25251507 - 25255526 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
