Assay Details
Gene Symbol: MIR2320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038612.1 | 1 | 41 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | centrosomal protein 104 | CEP104 | Sense | Chr. 6 - 65274092 - 65310735 on NCBI Build Sscrofa11.1 | ||
Downstream | DNA fragmentation factor subunit beta | DFFB | 9.5kb | Anti-sense | Chr. 6 - 65310868 - 65323816 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
