Assay Details
Gene Symbol: LOC100510918 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045041.1 | 3 | 2258 | 111 | ||
GenBank mRNA | AK235083.1 | - | 3 | 2258 | 111 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | TBC1 domain family member 1 | TBC1D1 | -247.1kb | Anti-sense | Chr. 8 - 29370344 - 29590574 on NCBI Build Sscrofa11.1 | |
Downstream | Kruppel like factor 3 | KLF3 | 185.5kb | Anti-sense | Chr. 8 - 30024978 - 30060981 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
