Assay Details
Gene Symbol: SLC50A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003125719.3 | XP_003125767.1 | 170 | 60 | ||
GenBank mRNA | AK398706.1 | - | 1 - 2 | 54 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
