Assay Details
| Gene Symbol: RACGAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091633.1 | XP_020947292.1 | 1972 | 56 | ||
| XM_021091634.1 | XP_020947293.1 | 1878 | 56 | |||
| XM_021091635.1 | XP_020947294.1 | 2005 | 56 | |||
| XM_021091636.1 | XP_020947295.1 | 1711 | 56 | |||
| XM_021091637.1 | XP_020947296.1 | 1737 | 56 | |||
| GenBank mRNA | AK236241.1 | - | 15 - 16 | 1910 | 56 | |
More Information
Set Membership: |
|

RACGAP1
Genomic Map