Assay Details
Gene Symbol: RSPO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001315656.1 | NP_001302585.1 | 2 - 3 | 295 | 111 | |
GenBank mRNA | JX910359.1 | - | 2 - 3 | 295 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
