Assay Details
| Gene Symbol: AP3S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021084652.1 | XP_020940311.1 | 193 | 64 | ||
| XM_021084653.1 | XP_020940312.1 | 193 | 64 | |||
| XM_021084654.1 | XP_020940313.1 | 518 | 64 | |||
| XM_021084656.1 | XP_020940315.1 | 160 | 64 | |||
| XM_021084657.1 | XP_020940316.1 | 202 | 64 | |||
| XM_021084658.1 | XP_020940317.1 | 193 | 64 | |||
| GenBank mRNA | AK237351.1 | - | 1 - 2 | 238 | 64 | |
| AK351449.1 | - | 1 - 2 | 147 | 64 | ||
| AK393965.1 | - | 1 - 2 | 162 | 64 | ||
| AY610105.1 | - | 1 - 2 | 150 | 64 | ||
More Information
Set Membership: |
|

AP3S1
Genomic Map