Assay Details
Gene Symbol: UTP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244742.1 | NP_001231671.1 | 1 - 2 | 75 | 100 | |
GenBank mRNA | AY609561.1 | - | 1 - 2 | 78 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
