Assay Details
Gene Symbol: UTP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244742.1 | NP_001231671.1 | 3 - 4 | 244 | 123 | |
GenBank mRNA | AY609561.1 | - | 3 - 4 | 247 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() |
