Assay Details
Gene Symbol: GEMIN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244865.1 | NP_001231794.1 | 3 - 4 | 240 | 70 | |
XM_005673429.3 | XP_005673486.1 | 186 | 70 | |||
XM_005673430.3 | XP_005673487.1 | 297 | 70 | |||
XM_013985910.2 | XP_013841364.1 | 303 | 70 | |||
XM_021079464.1 | XP_020935123.1 | 343 | 70 | |||
XM_021079465.1 | XP_020935124.1 | 172 | 70 | |||
XM_021079466.1 | XP_020935125.1 | 159 | 70 | |||
XM_021079467.1 | XP_020935126.1 | 136 | 70 | |||
GenBank mRNA | AK231675.1 | - | 3 - 4 | 240 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001244865.1, XM_005673429.3, XM_005673430.3, XM_013985910.2, XM_021079464.1, XM_021079465.1, XM_021079466.1, XM_021079467.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
