Assay Details
Gene Symbol: RNF141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252196.1 | NP_001239125.1 | 5 - 6 | 636 | 74 | |
XM_005661125.3 | XP_005661182.1 | 959 | 74 | |||
GenBank mRNA | AK344414.1 | - | 5 - 6 | 636 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
