Assay Details
| Gene Symbol: GPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257225.1 | NP_001244154.1 | 1 - 2 | 69 | 102 | |
| GenBank mRNA | HQ606483.1 | - | 1 - 2 | 69 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257225.1 transcripts.
Set Membership: |
|

GPR3
Genomic Map