Assay Details
Gene Symbol: FRRS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258382.2 | NP_001245311.2 | 1 - 2 | 343 | 60 | |
XM_021093620.1 | XP_020949279.1 | 306 | 60 | |||
XM_021093685.1 | XP_020949344.1 | 307 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
