Assay Details
Gene Symbol: C7H6orf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128447.2 | NP_001121919.2 | 1 - 2 | 81 | 114 | |
GenBank mRNA | HX231531.1 | - | 1 - 2 | 81 | 114 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001128447.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
