Assay Details
Gene Symbol: ZNF526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267848.1 | NP_001254777.1 | 1 - 2 | 33 | 66 | |
GenBank mRNA | AK233417.1 | - | 1 - 2 | 33 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001267848.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
