Assay Details
Gene Symbol: TNFAIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267890.1 | NP_001254819.1 | 1 - 2 | 299 | 55 | |
XM_005659182.3 | XP_005659239.1 | 665 | 55 | |||
XM_013992530.2 | XP_013847984.1 | 3768 | 55 | |||
XM_021077624.1 | XP_020933283.1 | 3738 | 55 | |||
GenBank mRNA | AB490119.1 | - | 1 - 2 | 299 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
