Assay Details
Gene Symbol: AKIRIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278752.1 | NP_001265681.1 | 1 - 2 | 240 | 107 | |
XM_021074590.1 | XP_020930249.1 | 840 | 107 | |||
GenBank mRNA | AK350524.1 | - | 1 - 2 | 823 | 107 | |
AK393334.1 | - | 1 - 2 | 814 | 107 | ||
AK395466.1 | - | 1 - 2 | 842 | 107 | ||
JN227885.1 | - | 1 - 2 | 240 | 107 | ||
KC140110.1 | - | 1 - 2 | 240 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
