Assay Details
Gene Symbol: TVP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281832.1 | NP_001268761.1 | 5 - 6 | 489 | 59 | |
XM_005653981.3 | XP_005654038.1 | 2043 | 59 | |||
XM_021066126.1 | XP_020921785.1 | 570 | 59 | |||
GenBank mRNA | AK230473.1 | - | 5 - 6 | 471 | 59 | |
AK389865.1 | - | 5 - 6 | 489 | 59 | ||
AY609392.1 | - | 5 - 6 | 471 | 59 | ||
BW959401.1 | - | 4 - 5 | 466 | 59 | ||
BW962415.1 | - | 5 - 6 | 489 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
