Assay Details
| Gene Symbol: KAT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284366.1 | NP_001271295.1 | 4 - 5 | 537 | 62 | |
| GenBank mRNA | AK235127.1 | - | 4 - 5 | 537 | 62 | |
| AK347664.1 | - | 4 - 5 | 863 | 62 | ||
| AK399133.1 | - | 4 - 5 | 525 | 62 | ||
| KF147893.1 | - | 4 - 5 | 537 | 62 | ||
More Information
Set Membership: |
|

KAT8
Genomic Map