Assay Details
Gene Symbol: ZFAND4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302672.1 | NP_001289601.1 | 8 - 9 | 2142 | 78 | |
XM_013983439.2 | XP_013838893.1 | 2278 | 78 | |||
XM_013983440.2 | XP_013838894.1 | 1992 | 78 | |||
XM_021071865.1 | XP_020927524.1 | 2135 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
