Assay Details
Gene Symbol: LOC100153094 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001315568.1 | NP_001302497.1 | 3 - 4 | 421 | 79 | |
GenBank mRNA | AK234724.1 | - | 3 - 4 | 411 | 79 | |
KP735787.1 | - | 3 - 4 | 421 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
