Assay Details
| Gene Symbol: EBI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315682.1 | NP_001302611.1 | 3 - 4 | 403 | 62 | |
| XM_005661336.3 | XP_005661393.1 | 1250 | 62 | |||
| GenBank mRNA | AB126827.1 | - | 3 - 4 | 404 | 62 | |
More Information
Set Membership: |
|

EBI3
Genomic Map