Assay Details
| Gene Symbol: SIGIRR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315689.1 | NP_001302618.1 | 4 - 5 | 623 | 65 | |
| GenBank mRNA | AB490122.1 | - | 4 - 5 | 623 | 65 | |
| AK235616.1 | - | 6 - 7 | 908 | 65 | ||
| AK239384.1 | - | 6 - 7 | 961 | 65 | ||
| AK396068.1 | - | 6 - 7 | 933 | 65 | ||
More Information
Set Membership: |
|

SIGIRR
Genomic Map