Assay Details
| Gene Symbol: LRP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315806.1 | NP_001302735.1 | 6 - 7 | 1711 | 69 | |
| XM_003125531.4 | XP_003125579.1 | 1770 | 69 | |||
| GenBank mRNA | JX092265.1 | - | 6 - 7 | 1711 | 69 | |
More Information
Set Membership: |
|

LRP12
Genomic Map