Assay Details
| Gene Symbol: CD8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348770.1 | NP_001335699.1 | 4 - 5 | 624 | 100 | |
| GenBank mRNA | AK238831.1 | - | 4 - 5 | 607 | 100 | |
| AK239664.1 | - | 4 - 5 | 605 | 100 | ||
| AK398365.1 | - | 4 - 5 | 613 | 100 | ||
More Information
Set Membership: |
|

CD8B
Genomic Map