Assay Details
| Gene Symbol: HEXB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213921.2 | NP_999086.2 | 12 - 13 | 1536 | 69 | |
| GenBank mRNA | AB529531.1 | - | 12 - 13 | 1447 | 69 | |
| AK233473.1 | - | 12 - 13 | 1536 | 69 | ||
| X92379.1 | - | 1458 | 69 | |||
More Information
Set Membership: |
|

HEXB
Genomic Map