Assay Details
| Gene Symbol: TGFB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214015.2 | NP_999180.2 | 5 - 6 | 1259 | 57 | |
| XM_021093503.1 | XP_020949162.1 | 1303 | 57 | |||
| GenBank mRNA | AF461808.1 | - | 5 - 6 | 856 | 57 | |
| AK344976.1 | - | 3 - 4 | 258 | 57 | ||
| M23703.1 | - | 5 - 6 | 1761 | 57 | ||
| X12373.1 | - | 1305 | 57 | |||
| Y00111.1 | - | 1259 | 57 | |||
More Information
Set Membership: |
|

TGFB1
Genomic Map