Assay Details
Gene Symbol: SLC25A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214418.2 | NP_999583.1 | 672 | 91 | ||
GenBank mRNA | AK231908.1 | - | 91 | |||
AK234164.1 | - | 91 | ||||
AK234290.1 | - | 91 | ||||
AK239239.1 | - | 91 | ||||
AK239241.1 | - | 91 | ||||
AK347345.1 | - | 91 | ||||
AK347755.1 | - | 91 | ||||
AK347916.1 | - | 91 | ||||
AK351616.1 | - | 91 | ||||
AK392846.1 | - | 91 | ||||
AK396724.1 | - | 91 | ||||
AK396746.1 | - | 91 | ||||
AK397157.1 | - | 91 | ||||
AK398428.1 | - | 91 | ||||
AK398628.1 | - | 91 | ||||
AK398826.1 | - | 91 | ||||
AK399005.1 | - | 91 | ||||
AK399036.1 | - | 91 | ||||
AK399165.1 | - | 91 | ||||
AK399729.1 | - | 91 | ||||
AK400296.1 | - | 91 | ||||
AK400766.1 | - | 91 | ||||
AK400849.1 | - | 91 | ||||
AK400977.1 | - | 91 | ||||
AY528245.2 | - | 672 | 91 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
