Assay Details
Gene Symbol: SLC25A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214418.2 | NP_999583.1 | 815 | 66 | ||
GenBank mRNA | AK231908.1 | - | 744 | 66 | ||
AK234164.1 | - | 776 | 66 | |||
AK234290.1 | - | 729 | 66 | |||
AK234558.1 | - | 840 | 66 | |||
AK239239.1 | - | 845 | 66 | |||
AK239241.1 | - | 806 | 66 | |||
AK347345.1 | - | 840 | 66 | |||
AK347755.1 | - | 742 | 66 | |||
AK347916.1 | - | 841 | 66 | |||
AK351616.1 | - | 824 | 66 | |||
AK352156.1 | - | 663 | 66 | |||
AK392846.1 | - | 843 | 66 | |||
AK396724.1 | - | 840 | 66 | |||
AK396746.1 | - | 66 | ||||
AK397157.1 | - | 844 | 66 | |||
AK398428.1 | - | 860 | 66 | |||
AK398628.1 | - | 840 | 66 | |||
AK398826.1 | - | 841 | 66 | |||
AK399005.1 | - | 838 | 66 | |||
AK399036.1 | - | 840 | 66 | |||
AK399165.1 | - | 843 | 66 | |||
AK399729.1 | - | 877 | 66 | |||
AK400296.1 | - | 840 | 66 | |||
AK400766.1 | - | 840 | 66 | |||
AK400849.1 | - | 66 | ||||
AK400977.1 | - | 840 | 66 | |||
AY528245.2 | - | 815 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
