Assay Details
| Gene Symbol: GSTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214389.2 | NP_999554.2 | 1 - 2 | 84 | 107 | |
| Gene Symbol: LOC106504562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | KP735782.1 | - | 1 - 2 | 344 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_214389.2 transcripts.
Set Membership: |
|

GSTA1
Genomic Map