Assay Details
| Gene Symbol: KLF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134344.2 | NP_001127816.1 | 1 - 2 | 156 | 103 | |
| GenBank mRNA | AK232376.1 | - | 1 - 2 | 170 | 103 | |
| AK400154.1 | - | 1 - 2 | 154 | 103 | ||
| EU669077.2 | - | 1 - 2 | 156 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001134344.2 transcripts.
Set Membership: |
|

KLF10
Genomic Map