Assay Details
| Gene Symbol: LOC102160336 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021071421.1 | XP_020927080.1 | 186 | 78 | ||
| GenBank mRNA | AK347921.1 | - | 1 - 2 | 129 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021071421.1 transcripts.
Set Membership: |
|

LOC102160336
Genomic Map