Assay Details
Gene Symbol: LOC102160336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021071421.1 | XP_020927080.1 | 553 | 125 | ||
GenBank mRNA | AK347921.1 | - | 4 - 5 | 496 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
