Assay Details
| Gene Symbol: LOC100738056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003482405.3 | XP_003482453.3 | 830 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100738056| Gene Symbol: LOC100738056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003482405.3 | XP_003482453.3 | 830 | 101 | ||
Set Membership: |
|