Assay Details
Gene Symbol: LRIT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005671160.1 | XP_005671217.1 | 110 | 108 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
